Novus Biologicals
Manufacturer Code:NB11074949
Catalog # NB11074949
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide as a part of rat and mouse transient receptor potential cation channel subfamily M member 4 (TRPM4) conjugated to immunogenic carrier protein was used as the immunogen. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Calcium-activated non-selective cation channel 1; Calcium-activated non-selective cation channel 1 FLJ20041 hTRPM4 Long transient receptor potential channel 4 LTrpC-4 LTRPC4 melastatin-4 PFHB1B transient receptor potential cation channel subfamily M member 4 transient receptor potential cation channel subfamily M member 4 TRPM4B; hTRPM4; Long transient receptor potential channel 4; LTrpC-4; Melastatin-4; Transient receptor potential cation channel subfamily M member 4; transient receptor potential cation channel, subfamily M, member 4
Gene Aliases: hTRPM4; LTRPC4; PFHB1B; TRPM4; TRPM4B
UniProt ID: (Human) A2RU25
Entrez Gene ID: (Human) 54795
Molecular Function: ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.