Invitrogen
Manufacturer Code:PA577327
Catalog # PIPA577327
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide KNKDDMPYMSQAQEIH(C) corresponding to amino acid residues 816-831 of human TRPM3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Long transient receptor potential channel 3; LTrpC-3; melastatin 2; Melastatin-2; MLSN2; Transient receptor potential cation channel subfamily M member 3; transient receptor potential melastatin 3 beta 1; transient receptor potential melastatin 3 beta 10; transient receptor potential melastatin 3 beta 11; transient receptor potential melastatin 3 beta 12; transient receptor potential melastatin 3 beta 13; transient receptor potential melastatin 3 beta 14; transient receptor potential melastatin 3 beta 15; transient receptor potential melastatin 3 beta 16; transient receptor potential melastatin 3 beta 17; transient receptor potential melastatin 3 beta 2; transient receptor potential melastatin 3 beta 3; transient receptor potential melastatin 3 beta 4; transient receptor potential melastatin 3 beta 5; transient receptor potential melastatin 3 beta 6; transient receptor potential melastatin 3 beta 7; transient receptor potential melastatin 3 beta 8; transient receptor potential melastatin 3 beta 9; TRPM3
Gene Aliases: 6330504P12Rik; 9330180E14; AU018608; B930001P07Rik; GON-2; KIAA1616; LTRPC3; MLSN2; TRPM3
UniProt ID: (Human) Q9HCF6
Entrez Gene ID: (Human) 80036, (Mouse) 226025, (Rat) 309407
Molecular Function:
ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.