Invitrogen
Manufacturer Code:OST00036W
Catalog # PIOST00036W
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide from the 1t extracellular loop of human TRPM3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: Long transient receptor potential channel 3; LTrpC-3; Melastatin-2; Melastatin-2 LTrpC3 MLSN2 GON-2; MLSN2; Transient receptor potential cation channel subfamily M member 3
Gene Aliases: GON-2; KIAA1616; LTRPC3; MLSN2; TRPM3
UniProt ID: (Human) Q9HCF6
Entrez Gene ID: (Human) 80036
Molecular Function:
ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.