Novus Biologicals
Manufacturer Code:NB100988640.025ML
Catalog # NB017958
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide from the 4th cytoplasmic loop of human TRPM3 conjugated to an immunogenic carrier protein was used as the immunogen. The antigen is homologous in rat mouse monkey and dog. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.3.1.31 EC 5.99.1.3 Long transient receptor potential channel 3 LTrpC3 LTrpC-3 LTRPC3KIAA1616MLSN2GON-2 melastatin 2 melastatin-2 transient receptor potential cation channel subfamily M member 3 transient receptor potential cation channel subfamily M member 3; Long transient receptor potential channel 3; LTrpC-3; Melastatin-2; MLSN2; Transient receptor potential cation channel subfamily M member 3
Gene Aliases: GON-2; KIAA1616; LTRPC3; MLSN2; TRPM3
UniProt ID: (Human) Q9HCF6
Entrez Gene ID: (Human) 80036
Molecular Function:
ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.