Invitrogen
Manufacturer Code:PA522882
Catalog # PIPA522882
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide made to an internal portion of the mouse protein (within residues 1200-1300). |
Conjugate | DyLight 650 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EREG1 KNP3 LTRPC2 NUDT9H NUDT9L1 TRPC7 LTrpC-2 estrogen-responsive element-associated gene 1 protein long transient receptor potential channel 2 transient receptor potential cation channel subfamily M member 2 transient receptor potential channel 7; Long transient receptor potential channel 2; LTrpC-2; null; Transient receptor potential cation channel subfamily M member 2; Transient receptor potential channel 7; transient receptor potential melastatin family 2; transient receptor protein 7; TrpC7
Gene Aliases: 9830168K16Rik; C79133; LTRPC2; Trp7; TRPC7; Trpm2; Trpm2-predicted; Trrp7
UniProt ID: (Rat) E9PTA2, (Mouse) Q91YD4
Entrez Gene ID: (Rat) 294329, (Mouse) 28240
Molecular Function:
ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.