Invitrogen
Manufacturer Code:OSR00186W
Catalog # PIOSR00186W
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide from aa region 750-800 of rat TRPM1 conjugated to an immunogenic carrier protein was used as the antigen |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Long transient receptor potential channel 1; LTrpC1; melastatin; Melastatin-1; Transient receptor potential cation channel subfamily M member 1; Transient receptor potential cation channel subfamily M member 1 variant delta1066-1071 Trpm1 LTRPC1 MLSN1 transient receptor potential cation c; trpm1 {ECO:0000312|EMBL:CAI30141.1}
Gene Aliases: 4732499L03Rik; AI606771; Ltrpc1; Mlsn1; Trpm1
UniProt ID: (Rat) Q2WEA5, (Mouse) Q2TV84
Entrez Gene ID: (Rat) 361586, (Mouse) 17364
Molecular Function: ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.