Proteintech
Manufacturer Code:551111AP150UL
Catalog # 50-173-5990
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Long transient receptor potential channel 1; LTrpC1; LTRPC1 Melastatin 1 MLSN MLSN1 TRPM1; Melastatin-1; Transient receptor potential cation channel subfamily M member 1; transient receptor potential cation channel, subfamily M, member 1; transient receptor potential melastatin family
Gene Aliases: CSNB1C; LTRPC1; MLSN; MLSN1; TRPM1
UniProt ID: (Human) F8W865
Entrez Gene ID: (Human) 4308
Molecular Function: ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.