Invitrogen
Manufacturer Code:PA529847
Catalog # PIPA529847
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 121 and 458 of Human TRPC6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: focal segmental glomerulosclerosis 2; FSGS2 TRP6 TRP-6 short transient receptor potential channel 6 transient receptor protein 6; Short transient receptor potential channel 6; transient receptor potential cation channel, subfamily C, member 6; Transient receptor protein 6; TRP-6; TrpC6
Gene Aliases: FSGS2; TRP6; TRPC6; Trrp6
UniProt ID: (Human) Q9Y210
Entrez Gene ID: (Human) 7225, (Rat) 89823
Molecular Function:
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.