Invitrogen
Manufacturer Code:PA129786
Catalog # PIPA129786
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide CRRNESQDYLLMDELG corresponding to N-term amino acids 24-38 of Mouse TRPC6. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C,Store at 4°C short term. For long term storage,avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Calcium entry channel; Short transient receptor potential channel 6; Transient receptor potential Ca2+ channel 6A trp6A transient receptor potential cation channel subfamily C member 6; Transient receptor protein 6; TRP-6; TrpC6
Gene Aliases: AV025995; LLHWJM002; LLHWJM003; LLHWJM004; mtrp6; TRP-6; Trp6; Trpc6; Trrp6
UniProt ID: (Mouse) Q61143
Entrez Gene ID: (Mouse) 22068
Molecular Function: ion channel transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.