Abnova
Manufacturer Code:PAB7504
Catalog # 89-133-313
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human TRPC6. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of TRPC6.
For Research Use Only
Protein Aliases: FLJ11098; FLJ14863; focal segmental glomerulosclerosis 2; FSGS2; Short transient receptor potential channel 6; transient receptor potential cation channel, subfamily C, member 6; Transient receptor protein 6; TRP-6; TRP6; TrpC6
Gene Aliases: FSGS2; TRP6; TRPC6
UniProt ID: (Human) Q9Y210
Entrez Gene ID: (Human) 7225
Molecular Function: ion channel transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.