Invitrogen
Manufacturer Code:PA580189
Catalog # PIPA580189
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human TRPC3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hTrp-3; ion channel protein; mTrp3; receptor-activated cation channel TRP3; Short transient receptor potential channel 3; Short transient receptor potential channel 3 TrpC3 Transient receptor protein 3 TRP-3 hTrp-3 hTrp3 TRPC3 TRP3; transient receptor potential cation channel subfamily C member 3; transient receptor potential cation channel subfamily C member 3 variant c; transient receptor potential cation channel, subfamily C, member 3; Transient receptor protein 3; TRP-3; trp-related protein 3; TrpC3
Gene Aliases: Mwk; SCA41; Trcp3; TRP3; TRPC3; TrpC3c; Trrp3
UniProt ID: (Human) Q13507
Entrez Gene ID: (Human) 7222, (Rat) 60395, (Mouse) 22065
Molecular Function:
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.