Invitrogen
Manufacturer Code:PA5116077
Catalog # PIPA5116077
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human TRPC3 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hTrp-3; hTrp3 hTrp-3 ion channel protein mTrp3 Mwk receptor-activated cation channel TRP3 SCA41 Short transient receptor potential channel 3 transient receptor potential cation channel subfamily C member 3 transient receptor potential cation channel subfamily C member 3 variant c transient receptor potential cation channel subfamily C member 3 transient receptor protein 3 Trcp3 TRP3 TRP-3 TRPC3 TrpC3c trp-related protein 3 Trrp3; ion channel protein; mTrp3; receptor-activated cation channel TRP3; Short transient receptor potential channel 3; transient receptor potential cation channel subfamily C member 3; transient receptor potential cation channel subfamily C member 3 variant c; transient receptor potential cation channel, subfamily C, member 3; Transient receptor protein 3; TRP-3; trp-related protein 3; TrpC3
Gene Aliases: Mwk; SCA41; Trcp3; TRP3; TRPC3; TrpC3c; Trrp3
UniProt ID: (Human) Q13507
Entrez Gene ID: (Human) 7222, (Rat) 60395, (Mouse) 22065
Molecular Function:
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.