Novus Biologicals
Manufacturer Code:NBP1703520.025ML
Catalog # NB018701
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide from the 4th cytoplasmic loop of rat TRPC3 conjugated to an immunogenic carrier protein was used as the antigen. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: hTrp-3; hTrp3 hTrp-3 short transient receptor potential channel 3 transient receptor potential cation channel subfamily C member 3 transient receptor potential channel 3 Transient receptor protein 3 TRP3 TRP-3 TrpC3; Short transient receptor potential channel 3; transient receptor potential cation channel subfamily C member 3 variant c; transient receptor potential cation channel, subfamily C, member 3; Transient receptor protein 3; TRP-3; TrpC3
Gene Aliases: SCA41; TRP3; TRPC3
UniProt ID: (Human) Q13507
Entrez Gene ID: (Human) 7222
Molecular Function:
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.