Abnova
Manufacturer Code:PAB14882
Catalog # 89-117-327
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus of mouse Trpc3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Trpc3.
For Research Use Only
Protein Aliases: MGC124333; mTrp3; receptor-activated cation channel TRP3; short transient receptor potential channel 3; transient receptor protein 3; Trcp3; TRP-3; trp-related protein 3; Trp3; Trrp3
Gene Aliases: Mwk; Trcp3; Trp3; Trrp3
Entrez Gene ID: (Mouse) 22065
Molecular Function:
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.