Abnova
Manufacturer Code:H00007220M03A
Catalog # 89-105-668
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1E4 |
Immunogen | TRPC1 (NP_003295442 a.a. &bsim 505 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant TRPC1.
For Research Use Only
Protein Aliases: capacitative calcium channel protein Trp1; HTRP-1; MGC133334; MGC133335; Short transient receptor potential channel 1; transient receptor potential canonical 1; transient receptor potential cation channel, subfamily C, member 1; Transient receptor protein 1; TRP-1; TRP1; TrpC1
Gene Aliases: HTRP-1; TRP1; TRPC1
UniProt ID: (Human) P48995
Entrez Gene ID: (Human) 7220
Molecular Function: ion channel transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.