Invitrogen
Manufacturer Code:PA5110525
Catalog # PIPA5110525
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 400-500 of human TRPC1 (NP_001238774.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3010009O07Rik AI115608 HTRP-1 membe Mtrp1 mTrp2 Short transient receptor potential channel 1 smTRPC2 Transient receptor protein 1 trp TRP1 TRP-1 TRP-1 protein trp2 TRPC1 TRPC2a TRPC2b Trp-related protein 1 Trrp1 Trrp2 XELAEV_18027727mg xtrpc1; capacitative calcium channel protein Trp1; Short transient receptor potential channel 1; transient receptor potential canonical 1; transient receptor potential cation channel, subfamily C, member 1; Transient receptor protein 1; TRP-1; Trp-related protein 1; TrpC1
Gene Aliases: HTRP-1; Mtrp1; TRP1; TRPC1; Trrp1
UniProt ID: (Human) P48995, (Mouse) Q61056
Entrez Gene ID: (Human) 7220, (Mouse) 22063
Molecular Function: ion channel transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.