Novus Biologicals
Manufacturer Code:NBP262000
Catalog # NBP262000
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-NLKQKRDENYQK from the internal region of the protein sequence according to NP_003295.1 NP_001238774.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
Protein Aliases: capacitative calcium channel protein Trp1; HTRP-1 MGC133334 MGC133335 transient receptor potential canonical 1 transient receptor potential cation channel subfamily C member 1 transient receptor potential channel 1 Transient receptor protein 1 TRP-1 TRP1short transient receptor potential channel 1 TrpC1; Short transient receptor potential channel 1; transient receptor potential canonical 1; transient receptor potential cation channel, subfamily C, member 1; Transient receptor protein 1; TRP-1; TrpC1
Gene Aliases: HTRP-1; TRP1; TRPC1
UniProt ID: (Human) P48995
Entrez Gene ID: (Human) 7220
Molecular Function:
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.