Proteintech
Manufacturer Code:194821AP150UL
Catalog # 50-173-2406
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: capacitative calcium channel protein Trp1; HTRP 1 Transient receptor protein 1 TRP 1 TRP1 TRPC1; Short transient receptor potential channel 1; transient receptor potential canonical 1; transient receptor potential cation channel, subfamily C, member 1; Transient receptor protein 1; TRP-1; Trp-related protein 1; TrpC1
Gene Aliases: HTRP-1; Mtrp1; TRP1; TRPC1; Trrp1
UniProt ID: (Human) P48995, (Mouse) Q61056, (Rat) Q9QX01
Entrez Gene ID: (Human) 7220, (Mouse) 22063, (Rat) 89821
Molecular Function:
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.