Invitrogen
Manufacturer Code:PA522835
Catalog # PIPA522835
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to a region within the N-terminus (residues 1-100) of the human TRPA1 protein. |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ANKTM1; ANKTM1 ankyrin-like with transmembrane domains 1 ankyrin-like with transmembrane domains protein 1 transformation-sensitive protein p120 transient receptor potential cation channel subfamily A member 1; ankyrin-like with transmembrane domains 1; Ankyrin-like with transmembrane domains protein 1; Transformation-sensitive protein p120; transient receptor potential A1; Transient receptor potential cation channel subfamily A member 1; transient receptor potential cation channel, subfamily A, member 1; Wasabi receptor
Gene Aliases: ANKTM1; FEPS; TRPA1; TRPA1b
UniProt ID: (Human) O75762, (Mouse) Q8BLA8
Entrez Gene ID: (Human) 8989, (Mouse) 277328, (Guinea pig) 100526649
Molecular Function: ion channel transporter voltage-gated ion channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.