Invitrogen
Manufacturer Code:OST00113W
Catalog # PIOST00113W
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide from the 1t extracellular loop of human TRPA1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ankyrin-like with transmembrane domains 1; Ankyrin-like with transmembrane domains protein 1; Transformation-sensitive protein p120; Transient receptor potential cation channel subfamily A member 1; Transient receptor potential cation channel subfamily A member 1 ankyrin-like with transmembrane domains protein 1 transformation sensitive protein p120 ANKTM1; transient receptor potential cation channel, subfamily A, member 1; Wasabi receptor
Gene Aliases: ANKTM1; FEPS; TRPA1
UniProt ID: (Human) O75762
Entrez Gene ID: (Human) 8989
Molecular Function:
ion channel
transporter
voltage-gated ion channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.