Novus Biologicals
Manufacturer Code:NB100988410.025ML
Catalog # NB017955
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide from the 1t cytoplasmic loop of rat mouse TRPA1 conjugated to an immunogenic carrier protein was used as the immunogen. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ANKTM1ankyrin-like with transmembrane domains 1 Ankyrin-like with transmembrane domains protein 1 Transformation-sensitive protein p120 transient receptor potential cation channel subfamily A member 1 transient receptor potential cation channel subfamily A member 1; ankyrin-like with transmembrane domains 1; Ankyrin-like with transmembrane domains protein 1; Transformation-sensitive protein p120; Transient receptor potential cation channel subfamily A member 1; transient receptor potential cation channel, subfamily A, member 1; Wasabi receptor
Gene Aliases: ANKTM1; FEPS; TRPA1
UniProt ID: (Human) O75762
Entrez Gene ID: (Human) 8989
Molecular Function:
ion channel
transporter
voltage-gated ion channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.