Invitrogen
Manufacturer Code:PA541792
Catalog # PIPA541792
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human TROVE2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 60 kDa ribonucleoprotein Ro; 60 kDa Ro protein; 60 kDa SS-A/Ro ribonucleoprotein; 60 kDa SS-A/Ro ribonucleoprotein-like protein; gastric cancer multi-drug resistance protein; Ro 60 kDa autoantigen; Ro60 autoantigen; roRNP; Sjoegren syndrome antigen A2; Sjoegren syndrome type A antigen; Sjogren syndrome antigen A2; Sjogren syndrome antigen A2 (60kD, ribonucleoprotein autoantigen SS-A/Ro); SS-A; TROVE domain family member 2; TROVE domain family, member 2
Gene Aliases: 1810007I17Rik; A530054J02Rik; AI646302; RO60; RORNP; SS-A/Ro; Ssa; SSA2; TROVE2
UniProt ID: (Human) P10155, (Mouse) O08848
Entrez Gene ID: (Rabbit) 100348767, (Horse) 100051140, (Dog) 478957, (Bovine) 518830, (Guinea pig) 100714507, (Human) 6738, (Mouse) 20822, (Rat) 304833
Molecular Function:
RNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.