Invitrogen
Manufacturer Code:PA5110661
Catalog # PIPA5110661
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human TROVE2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1810007I17Rik 60 kDa Ro protein 60 kDa SS-A/Ro ribonucleoprotein A530054J02Rik AI646302 BOS_15617 Ro 60 kDa autoantigen RO60 Ro60 autoantigen RORNP RP11-101E13.3 Sjoegren syndrome antigen A2 Sjoegren syndrome type A antigen Ssa SS-A SS-A/Ro SSA2 TROVE domain family member 2 TROVE2; 60 kDa ribonucleoprotein Ro; 60 kDa Ro protein; 60 kDa SS-A/Ro ribonucleoprotein; gastric cancer multi-drug resistance protein; Ro 60 kDa autoantigen; Ro60 autoantigen; Sjoegren syndrome antigen A2; Sjoegren syndrome type A antigen; Sjogren syndrome antigen A2 (60kD, ribonucleoprotein autoantigen SS-A/Ro); SS-A; TROVE domain family member 2; TROVE domain family, member 2
Gene Aliases: RO60; RORNP; SSA2; TROVE2
UniProt ID: (Human) P10155
Entrez Gene ID: (Human) 6738
Molecular Function: RNA binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.