Invitrogen
Manufacturer Code:PA598663
Catalog # PIPA598663
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human CCA tRNA nucleotidyltransferase 1 mitochondrial protein (128-293AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP(CTP):tRNA nucleotidyltransferase; CCA tRNA nucleotidyltransferase 1, mitochondrial; CCA-adding enzyme; mitochondrial CCA-adding tRNA-nucleotidyltransferase; Mitochondrial tRNA nucleotidyl transferase, CCA-adding; mt CCA-adding enzyme; mt tRNA adenylyltransferase; mt tRNA CCA-diphosphorylase; mt tRNA CCA-pyrophosphorylase; tRNA CCA nucleotidyl transferase 1; tRNA nucleotidyl transferase, CCA-adding, 1; tRNA-nucleotidyltransferase 1, mitochondrial
Gene Aliases: CCA1; CGI-47; MtCCA; RPEM; SIFD; TRNT1
UniProt ID: (Human) Q96Q11
Entrez Gene ID: (Human) 51095
Molecular Function:
RNA binding protein
mRNA polyadenylation factor
mRNA processing factor
nucleic acid binding
nucleotidyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.