Invitrogen
Manufacturer Code:PA541859
Catalog # PIPA541859
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human TRNT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP(CTP):tRNA nucleotidyltransferase; CCA tRNA nucleotidyltransferase 1, mitochondrial; CCA-adding enzyme; mitochondrial CCA-adding tRNA-nucleotidyltransferase; Mitochondrial tRNA nucleotidyl transferase, CCA-adding; mt CCA-adding enzyme; mt tRNA adenylyltransferase; mt tRNA CCA-diphosphorylase; mt tRNA CCA-pyrophosphorylase; tRNA adenylyltransferase, mitochondrial; tRNA CCA nucleotidyl transferase 1; tRNA nucleotidyl transferase, CCA-adding, 1; tRNA-nucleotidyltransferase; tRNA-nucleotidyltransferase 1 mitochondrial-like protein; tRNA-nucleotidyltransferase 1, mitochondrial
Gene Aliases: 2410043H24Rik; 2610044E04Rik; 9830143O18Rik; AA408384; bMtCCA; C76540; CCA1; CGI-47; mt-Trat; MtCCA; RPEM; SIFD; TRNT1; zgc:86645
UniProt ID: (Human) Q96Q11, (Mouse) Q8K1J6
Entrez Gene ID: (Human) 51095, (Dog) 476552, (Horse) 100053009, (Bovine) 407144, (Rabbit) 100347348, (Guinea pig) 100735013, (Rat) 312616, (Mouse) 70047, (Zebrafish) 415249
Molecular Function: RNA binding protein mRNA polyadenylation factor mRNA processing factor nucleic acid binding nucleotidyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.