Proteintech
Manufacturer Code:150531AP150UL
Catalog # 50-172-9935
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TRNAU1APSECP43 Fusion Protein Ag7036 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PRO1902 RP4 669K10.4 SECP43 TRNAU1AP TRSPAP1; SECp43; tRNA selenocysteine 1-associated protein 1; tRNA selenocysteine associated protein; tRNA selenocysteine associated protein (SECP43); tRNA selenocysteine associated protein 1; tRNA selenocysteine-associated protein 1
Gene Aliases: 1110007F05Rik; AA387999; PRO1902; SECP43; TRNAU1AP; TRSPAP1; WI1-2643G14.2
UniProt ID: (Human) Q9NX07, (Mouse) Q80VC6, (Rat) Q9QZI7
Entrez Gene ID: (Human) 54952, (Mouse) 71787, (Rat) 65241
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.