Abnova
Manufacturer Code:H00054952K
Catalog # 89-207-684
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human TRNAU1AP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human TRNAU1AP peptide using ARM Technology.
For Research Use Only
Protein Aliases: FLJ20503 PRO1902 RP4-669K10.4 SECP43 TRSPAP1; SECp43; tRNA selenocysteine 1-associated protein 1; tRNA selenocysteine associated protein (SECP43); tRNA selenocysteine associated protein 1; tRNA selenocysteine-associated protein 1
Gene Aliases: PRO1902; SECP43; TRNAU1AP; TRSPAP1
UniProt ID: (Human) Q9NX07
Entrez Gene ID: (Human) 54952
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.