OriGene
Manufacturer Code:TA505699
Catalog # 50-167-6136
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2A4 |
Immunogen | Full length human recombit protein of human TRMU produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: LCAL3 MTO2 MTU1 TRMT TRMT1; lung cancer associated lncRNA 3; mitochondrial 5-methylaminomethyl-2-thiouridylate-methyltransferase; Mitochondrial tRNA-specific 2-thiouridylase 1; MTO2 homolog; tRNA (5-methylaminomethyl-2-thiouridylate)-methyltransferase 1; tRNA 5-methylaminomethyl-2-thiouridylate methyltransferase
Gene Aliases: 1110005N20Rik; 1600025P05Rik; AI314320; LCAL3; MTO2; MTU1; TRMT; TRMT1; TRMU; TRNT1
UniProt ID: (Human) O75648, (Mouse) Q9DAT5
Entrez Gene ID: (Human) 55687, (Dog) 481204, (Mouse) 72026
Molecular Function:
RNA binding protein
RNA methyltransferase
methyltransferase
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.