OriGene
Manufacturer Code:TA505698
Catalog # 50-167-6135
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1C6 |
Immunogen | Full length human recombit protein of human TRMU produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: LCAL3 MTO2 MTU1 TRMT TRMT1; lung cancer associated lncRNA 3; mitochondrial 5-methylaminomethyl-2-thiouridylate-methyltransferase; Mitochondrial tRNA-specific 2-thiouridylase 1; MTO2 homolog
Gene Aliases: LCAL3; MTO2; MTU1; TRMT; TRMT1; TRMU; TRNT1
UniProt ID: (Human) O75648
Entrez Gene ID: (Human) 55687
Molecular Function: RNA binding protein RNA methyltransferase methyltransferase nucleic acid binding transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.