Novus Biologicals
Manufacturer Code:NBP30322620UL
Catalog # NB095399
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 110-330 of human TRMT9B (NP_065895.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C8orf79 EC 2.1.1.- KIAA1456; Probable tRNA methyltransferase 9-like protein; Probable tRNA methyltransferase 9B
Gene Aliases: C8orf79; KIAA1456; TRM9L; TRMT9B
UniProt ID: (Human) Q9P272
Entrez Gene ID: (Human) 57604
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.