Invitrogen
Manufacturer Code:PA555406
Catalog # PIPA555406
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human TRMT61B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: mRNA methyladenosine-N(1)-methyltransferase; potential tRNA (adenine(58)-N(1))-methyltransferase catalytic subunit TRMT61B; potential tRNA (adenine-N(1)-)-methyltransferase catalytic subunit TRMT61B; tRNA (adenine(58)-N(1))-methyltransferase, mitochondrial; tRNA methyltransferase 61 homolog B
Gene Aliases: TRMT61B
UniProt ID: (Human) Q9BVS5
Entrez Gene ID: (Human) 55006
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.