Novus Biologicals
Manufacturer Code:NBP183640
Catalog # NBP183640
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:GVCAVYVVNITQVIELLDGIRTCELALSCEKISEVIVRDWLVCLAKQKNGILAQKVESKINTDVQLDSQEKIGVKGELFQEDDHEESHSDFPYGSFPYVARPVHWQPGHTAFLVKLRKV |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp564I2178 EC 2.1.1.36 FLJ20628 potential tRNA (adenine-N(1)-)-methyltransferase catalytic subunit TRMT61B tRNA methyltransferase 61 homolog B (S. cerevisiae); mRNA methyladenosine-N(1)-methyltransferase; potential tRNA (adenine(58)-N(1))-methyltransferase catalytic subunit TRMT61B; potential tRNA (adenine-N(1)-)-methyltransferase catalytic subunit TRMT61B; tRNA (adenine(58)-N(1))-methyltransferase, mitochondrial; tRNA methyltransferase 61 homolog B
Gene Aliases: TRMT61B
UniProt ID: (Human) Q9BVS5
Entrez Gene ID: (Human) 55006
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.