Invitrogen
Manufacturer Code:MA527125
Catalog # PIMA527125
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1B5 |
Immunogen | Human recombit protein fragment corresponding to amino acids 1-303 of TRMT5 produced in E.coli |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: M1G-methyltransferase; TRM5 tRNA methyltransferase 5 homolog; TRMT5; tRNA (guanine(37)-N1)-methyltransferase; tRNA (guanine-N(1)-)-methyltransferase; tRNA (guanine-N(1)-)-methyltransferase M1G-methyltransferase tRNA &lsqbGM37] methyltransferase tRNA methyltransferase 5 TRM5 KIAA1393 TRMT5; tRNA [GM37] methyltransferase; tRNA methyltransferase 5 homolog; tRNA-N1G37 methyltransferase
Gene Aliases: COXPD26; KIAA1393; TRM5; TRMT5
UniProt ID: (Human) Q32P41
Entrez Gene ID: (Human) 57570
Molecular Function:
methyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.