Invitrogen
Manufacturer Code:PA551488
Catalog # PIPA551488
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human TRMT5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: M1G-methyltransferase; TRM5 tRNA methyltransferase 5 homolog; TRMT5; tRNA (guanine(37)-N1)-methyltransferase; tRNA (guanine-N(1)-)-methyltransferase; tRNA (guanine-N(1)-)-methyltransferase M1G-methyltransferase tRNA &lsqbGM37] methyltransferase tRNA methyltransferase 5 TRM5 KIAA1393 TRMT5; tRNA [GM37] methyltransferase; tRNA methyltransferase 5 homolog; tRNA-N1G37 methyltransferase
Gene Aliases: COXPD26; KIAA1393; TRM5; TRMT5
UniProt ID: (Human) Q32P41
Entrez Gene ID: (Human) 57570
Molecular Function: methyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.