Invitrogen
Manufacturer Code:PA5113880
Catalog # PIPA5113880
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human TRMT5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2610027O18Rik BOS_11048 COXPD26 KIAA1393 LOC100343165 M1G-methyltransferase mKIAA1393 RGD1306567 Trm5 TRM5 tRNA methyltransferase 5 TRM5 tRNA methyltransferase 5 homolog TRM5 tRNA methyltransferase 5 homolog (S. cerevisiae) TRMT5 tRNA (guanine(37)-N1)-methyltransferase tRNA (guanine-N(1)-)-methyltransferase tRNA (guanine-N(1)-)-methyltransferase-like tRNA [GM37] methyltransferase tRNA methyltransferase 5 tRNA methyltransferase 5 homolog tRNA-N1G37 methyltransferase; M1G-methyltransferase; TRM5 tRNA methyltransferase 5 homolog; TRMT5; tRNA (guanine(37)-N1)-methyltransferase; tRNA (guanine-N(1)-)-methyltransferase; tRNA [GM37] methyltransferase; tRNA methyltransferase 5 homolog; tRNA-N1G37 methyltransferase
Gene Aliases: COXPD26; KIAA1393; TRM5; TRMT5
UniProt ID: (Human) Q32P41
Entrez Gene ID: (Human) 57570
Molecular Function:
methyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.