Proteintech
Manufacturer Code:182551AP150UL
Catalog # 50-173-2072
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TRMT5 Fusion Protein Ag13075 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA1393 M1G methyltransferase TRM5 TRMT5 tRNA &lsqbGM37] methyltransferase tRNA methyltransferase 5; M1G-methyltransferase; TRM5 tRNA methyltransferase 5 homolog; TRMT5; tRNA (guanine(37)-N1)-methyltransferase; tRNA (guanine-N(1)-)-methyltransferase; tRNA [GM37] methyltransferase; tRNA methyltransferase 5 homolog; tRNA-N1G37 methyltransferase
Gene Aliases: 2610027O18Rik; COXPD26; KIAA1393; mKIAA1393; RGD1306567; TRM5; TRMT5
UniProt ID: (Human) Q32P41, (Mouse) Q9D0C4
Entrez Gene ID: (Human) 57570, (Mouse) 76357, (Rat) 362754
Molecular Function:
methyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.