Proteintech
Manufacturer Code:207671AP150UL
Catalog # 50-173-3070
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TRMT2B Fusion Protein Ag14750 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CXorf34 TRM2 homolog TRMT2B; TRM2 homolog; TRM2 tRNA methyltransferase 2 homolog B; tRNA (uracil(54)-C(5))-methyltransferase homolog; tRNA (uracil-5-)-methyltransferase homolog
Gene Aliases: CXorf34; dJ341D10.3; TRMT2B
UniProt ID: (Human) Q96HU7
Entrez Gene ID: (Human) 79979
Molecular Function:
RNA binding protein
RNA methyltransferase
methyltransferase
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.