Invitrogen
Manufacturer Code:PA599786
Catalog # PIPA599786
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human TRMT11 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2410075D05Rik 3110045I18Rik AW213713 C6orf75 MDS024 putative RNA methylase MDS024 TRM11 Trmt11 TRMT11-1 tRNA (guanine(10)-N2)-methyltransferase homolog tRNA guanosine-2'-O-methyltransferase TRM11 homolog tRNA methyltransferase 11 tRNA methyltransferase 11 homolog tRNA methyltransferase 11 homolog (S. cerevisiae); tRNA (guanine(10)-N2)-methyltransferase homolog; tRNA guanosine-2'-O-methyltransferase TRM11 homolog; tRNA methyltransferase 11 homolog
Gene Aliases: 2410075D05Rik; 3110045I18Rik; AW213713; C6orf75; MDS024; TRM11; TRMT11; TRMT11-1
UniProt ID: (Human) Q7Z4G4
Entrez Gene ID: (Human) 60487, (Mouse) 73681
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.