Invitrogen
Manufacturer Code:PA567688
Catalog # PIPA567688
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human TRMT11. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C6orf75; tRNA (guanine(10)-N2)-methyltransferase homolog; tRNA guanosine-2'-O-methyltransferase TRM11 homolog
Gene Aliases: C6orf75; MDS024; TRM11; TRMT11; TRMT11-1
UniProt ID: (Human) Q7Z4G4
Entrez Gene ID: (Human) 60487
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.