Invitrogen
Manufacturer Code:PA543190
Catalog # PIPA543190
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human TRMT11 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: putative RNA methylase MDS024; tRNA (guanine(10)-N2)-methyltransferase homolog; tRNA guanosine-2'-O-methyltransferase TRM11 homolog; tRNA guanosine-2'-O-methyltransferase TRM11-like protein-like protein; tRNA methyltransferase 11 homolog
Gene Aliases: 2410075D05Rik; 3110045I18Rik; AW213713; C6orf75; MDS024; TRM11; TRMT11; TRMT11-1
UniProt ID: (Bovine) Q05B63, (Human) Q7Z4G4, (Rat) Q7TNK6
Entrez Gene ID: (Bovine) 514867, (Dog) 476285, (Horse) 100073022, (Rabbit) 100338244, (Human) 60487, (Guinea pig) 100728385, (Mouse) 73681, (Rat) 378794
Molecular Function:
RNA binding protein
RNA methyltransferase
methyltransferase
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.