Proteintech
Manufacturer Code:602051IG150UL
Catalog # 50-173-6307
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5D8C7 |
Immunogen | TRIP6 Fusion Protein Ag15149 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: OIP 1 OIP1 Opa interacting protein 1 TR interacting protein 6 TRIP 6 TRIP6 ZRP 1 Zyxin related protein 1; OIP-1; OPA-interacting protein 1; thyroid hormone receptor interacting protein 6; Thyroid receptor-interacting protein 6; TR-interacting protein 6; TRIP-6; ZRP-1; zyxin related protein 1; Zyxin-related protein 1
Gene Aliases: OIP-1; OIP1; TRIP-6; TRIP6; TRIP6i2; ZRP-1; Zrp1
UniProt ID: (Human) Q15654, (Mouse) Q9Z1Y4
Entrez Gene ID: (Human) 7205, (Mouse) 22051
Molecular Function:
actin family cytoskeletal protein
cell junction protein
cytoskeletal protein
enzyme modulator
kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.