Invitrogen
Manufacturer Code:PA530723
Catalog # PIPA530723
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 1 and 443 of Human TRIP15 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ALIEN CSN2 SGN2 TRIP15 COP9 signalosome complex subunit 2 JAB1-containing signalosome subunit 2 TR-interacting protein 15 TRIP-15 alien homolog signalosome subunit 2 thyroid receptor interacting protein 15 thyroid receptor-interacting protein 15; Alien homolog; alien-like; COP9 constitutive photomorphogenic homolog subunit 2; COP9 signalosome complex subunit 2; JAB1-containing signalosome subunit 2; SGN2; signalosome subunit 2; thyroid receptor interacting protein 15; Thyroid receptor-interacting protein 15; TR-interacting protein 15; TRIP-15
Gene Aliases: AI315723; ALIEN; C85265; COPS2; CSN2; SGN2; TRIP-15; TRIP15
UniProt ID: (Human) P61201, (Mouse) P61202
Entrez Gene ID: (Human) 9318, (Mouse) 12848
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.