Invitrogen
Manufacturer Code:PA518161
Catalog # PIPA518161
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (LNSLNQAVVSKLA) corresponding to the C-terminus amino acids of COPS2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Alien homolog; alien-like; COP9 constitutive photomorphogenic homolog subunit 2; COP9 signalosome complex subunit 2; COPS2 TRIP15 COPS2 CSN2 thyroid receptor interacting protein 15 SGN2 COP9 constitutive photomorphogenic homolog subunit 2 thyroid receptor interacting protein 15 COP9 constitutive photomorphogenic homolog subunit 2 (Arabidopsis) COP9 subunit 2 ALIEN COP9 signalosome complex subunit 2 JAB1-containing signalosome subunit 2; JAB1-containing signalosome subunit 2; SGN2; signalosome subunit 2; thyroid receptor interacting protein 15; Thyroid receptor-interacting protein 15; TR-interacting protein 15; TRIP-15
Gene Aliases: AI315723; ALIEN; C85265; COPS2; CSN2; SGN2; TRIP-15; TRIP15
UniProt ID: (Human) P61201, (Mouse) P61202
Entrez Gene ID: (Human) 9318, (Mouse) 12848
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.