Invitrogen
Manufacturer Code:PA557840
Catalog # PIPA557840
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human TRIP12 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase for Arf; E3 ubiquitin-protein ligase TRIP12; HECT-type E3 ubiquitin transferase TRIP12; probable E3 ubiquitin-protein ligase TRIP12; thyroid receptor interacting protein 12; Thyroid receptor-interacting protein 12; TR-interacting protein 12; ULF; ULF TRIP-12 KIAA0045 ULF E3 ubiquitin-protein ligase TRIP12 E3 ubiquitin-protein ligase for Arf
Gene Aliases: KIAA0045; TRIP-12; TRIP12; ULF
UniProt ID: (Human) Q14669
Entrez Gene ID: (Human) 9320
Molecular Function:
ligase
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.