Invitrogen
Manufacturer Code:PA538016
Catalog # PIPA538016
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | 18 amino acid peptide near the carboxy terminus of human TRIP12 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase for Arf; E3 ubiquitin-protein ligase TRIP12; gene trap locus 6; HECT-type E3 ubiquitin transferase TRIP12; probable E3 ubiquitin-protein ligase TRIP12; thyroid receptor interacting protein 12; thyroid receptor interactor TRIP12; Thyroid receptor-interacting protein 12; TR-interacting protein 12; ULF; ULF TRIP-12 KIAA0045 ULF E3 ubiquitin-protein ligase TRIP12 E3 ubiquitin-protein ligase for Arf
Gene Aliases: 1110036I07Rik; 6720416K24Rik; AA410158; Gtl6; KIAA0045; TRIP-12; TRIP12; ULF
UniProt ID: (Human) Q14669, (Mouse) G5E870, (Rat) F1LP64
Entrez Gene ID: (Human) 9320, (Mouse) 14897, (Rat) 316575
Molecular Function:
ligase
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.