Novus Biologicals
Manufacturer Code:NBP293144100UL
Catalog # NB028176
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 180-310 of human TRIOBP (NP_008963.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ39315 TARA TRIO and F-actin binding protein TRIO and F-actin-binding protein; Protein Tara; tara-like protein; TRIO and F-actin-binding protein; Trio-associated repeat on actin
Gene Aliases: DFNB28; dJ37E16.4; HRIHFB2122; KIAA1662; TAP68; TARA; TRIOBP
UniProt ID: (Human) Q9H2D6
Entrez Gene ID: (Human) 11078
Molecular Function: G-protein modulator actin family cytoskeletal protein cytoskeletal protein enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.