Abnova
Manufacturer Code:PAB21237
Catalog # 89-122-729
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human TRIOBP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant TRIOBP.
For Research Use Only
Protein Aliases: DFNB28; dJ37E16.4; FLJ39315; HRIHFB2122; KIAA1662; Protein Tara; TARA; tara-like protein; TRIO and F-actin-binding protein; Trio-associated repeat on actin
Gene Aliases: DFNB28; dJ37E16.4; HRIHFB2122; KIAA1662; TAP68; TARA; TRIOBP
UniProt ID: (Human) Q9H2D6
Entrez Gene ID: (Human) 11078
Molecular Function:
G-protein modulator
actin family cytoskeletal protein
cytoskeletal protein
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.