Invitrogen
Manufacturer Code:PA580188
Catalog # PIPA580188
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human TRIO. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PTPRF-interacting protein; triple functional domain (PTPRF interacting); Triple functional domain protein; Triple functional domain protein PTPRF-interacting protein TRIO
Gene Aliases: 6720464I07Rik; AA408740; ARHGEF23; Solo; tgat; TRIO
UniProt ID: (Human) O75962, (Mouse) Q0KL02, (Rat) F1M0Z1
Entrez Gene ID: (Human) 7204, (Mouse) 223435, (Rat) 310192
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.