Invitrogen
Manufacturer Code:PA572660
Catalog # PIPA572660
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Zebrafish |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 2310-2344 amino acids from the Central region of human (DANRE) trio |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARHGEF23 MEBAS MRD44 tgat triple functional domain protein PTPRF-interacting protein triple functional domain (PTPRF interacting); triple functional domain (PTPRF interacting)
Gene Aliases: si:ch211-13c6.1; si:dkey-158b13.2; trio
Entrez Gene ID: (Zebrafish) 557667
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.