Novus Biologicals
Manufacturer Code:NBP298074AF488
Catalog # NB052142
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human TRIO fragment. |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARHGEF23 EC 2.7.11.1 FLJ42780 PTPRF-interacting protein tgat triple functional domain (PTPRF interacting) triple functional domain protein; PTPRF-interacting protein; triple functional domain (PTPRF interacting); Triple functional domain protein
Gene Aliases: ARHGEF23; tgat; TRIO
UniProt ID: (Human) O75962
Entrez Gene ID: (Human) 7204
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.